Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597105.2 | XP_005597162.1 | 1218 | 73 | ||
XM_005597106.3 | XP_005597163.1 | 1805 | 73 | |||
XM_005597107.3 | XP_005597164.1 | 1337 | 73 | |||
XM_005597110.3 | XP_005597167.1 | 683 | 73 | |||
XM_023651940.1 | XP_023507708.1 | 1836 | 73 | |||
XM_023651941.1 | XP_023507709.1 | 1520 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
