Assay Details
| Gene Symbol: JPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492365.6 | XP_001492415.2 | 341 | 71 | ||
| XM_003362502.4 | XP_003362550.1 | 341 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: JPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492365.6 | XP_001492415.2 | 341 | 71 | ||
| XM_003362502.4 | XP_003362550.1 | 341 | 71 | |||
Set Membership: |
|