Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362744.4 | XP_003362792.2 | 304 | 91 | ||
| XM_023615996.1 | XP_023471764.1 | 313 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF771| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362744.4 | XP_003362792.2 | 304 | 91 | ||
| XM_023615996.1 | XP_023471764.1 | 313 | 91 | |||
Set Membership: |
|