Assay Details
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497464.5 | XP_001497514.2 | 157 | 99 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497464.5 | XP_001497514.2 | 157 | 99 |