Assay Details
| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635564.1 | XP_023491332.1 | 773 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDLRAD2| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635564.1 | XP_023491332.1 | 773 | 95 | ||