Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636000.1 | XP_023491768.1 | 996 | 72 | ||
| XM_023636001.1 | XP_023491769.1 | 1129 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636000.1 | XP_023491768.1 | 996 | 72 | ||
| XM_023636001.1 | XP_023491769.1 | 1129 | 72 | |||
Set Membership: |
|