Assay Details
| Gene Symbol: LRFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654532.1 | XP_023510300.1 | 4062 | 109 | ||
| XM_023654533.1 | XP_023510301.1 | 4376 | 109 | |||
More Information
Set Membership: |
|

LRFN4
Genomic Map