Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653675.1 | XP_023509443.1 | 5417 | 126 | ||
| XM_023653676.1 | XP_023509444.1 | 1555 | 126 | |||
More Information
Set Membership: |
|

RASD1
Genomic Map