Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245977.1 | NP_001232906.1 | 2 - 3 | 369 | 54 | |
| XM_023631009.1 | XP_023486777.1 | 2119 | 54 | |||
| XM_023631010.1 | XP_023486778.1 | 2037 | 54 | |||
| GenBank mRNA | JF502272.1 | - | 2 - 3 | 369 | 54 | |
More Information
Set Membership: |
|

TNFRSF13C
Genomic Map