Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417496.6 | XP_417496.5 | 1 - 2 | 907 | 60 | |
| GenBank mRNA | AF205877.1 | - | 1 - 2 | 320 | 60 | |
| S77477.1 | - | 1 - 2 | 66 | 60 | ||
More Information
Set Membership: |
|

BMP7
Genomic Map