Assay Details
| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297422.2 | XP_015152908.1 | 2 - 3 | 1897 | 65 | |
| GenBank mRNA | AF486810.1 | - | 1 - 2 | 387 | 65 | |
More Information
Set Membership: |
|

NODAL
Genomic Map