Assay Details
Gene Symbol: SLC25A21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287659.2 | XP_015143145.1 | 4 - 5 | 361 | 94 | |
XM_025150917.1 | XP_025006685.1 | 5 - 6 | 512 | 94 | ||
XM_025150918.1 | XP_025006686.1 | 5 - 6 | 512 | 94 | ||
XM_025150919.1 | XP_025006687.1 | 5 - 6 | 512 | 94 | ||
XM_025150920.1 | XP_025006688.1 | 4 - 5 | 428 | 94 | ||
XM_025150921.1 | XP_025006689.1 | 4 - 5 | 386 | 94 | ||
XM_025150922.1 | XP_025006690.1 | 4 - 5 | 237 | 94 | ||
XM_025150923.1 | XP_025006691.1 | 3 - 4 | 311 | 94 | ||
GenBank mRNA | BX931882.1 | - | 4 - 5 | 287 | 94 | |
BX935431.2 | - | 4 - 5 | 263 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
