Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281668.2 | XP_015137154.1 | 3 - 4 | 987 | 76 | |
| XM_015281669.2 | XP_015137155.1 | 3 - 4 | 703 | 76 | ||
| GenBank mRNA | BX932603.1 | - | 1 - 2 | 107 | 76 | |
More Information
Set Membership: |
|

KBTBD2
Genomic Map