Assay Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199473.1 | NP_001186402.1 | 2 - 3 | 226 | 67 | |
| GenBank mRNA | BX930666.2 | - | 2 - 3 | 209 | 67 | |
| BX931460.2 | - | 2 - 3 | 209 | 67 | ||
| BX932279.2 | - | 2 - 3 | 209 | 67 | ||
| BX932726.1 | - | 2 - 3 | 219 | 67 | ||
| CN228366.1 | - | 2 - 3 | 209 | 67 | ||
| CR353353.1 | - | 2 - 3 | 215 | 67 | ||
| CR406236.1 | - | 2 - 3 | 210 | 67 | ||
More Information
Set Membership: |
|

MRPL53
Genomic Map