Assay Details
| Gene Symbol: PSMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198602.1 | NP_001185531.1 | 6 - 7 | 496 | 67 | |
| XM_015278878.2 | XP_015134364.1 | 6 - 7 | 517 | 67 | ||
| GenBank mRNA | BX933615.1 | - | 6 - 7 | 513 | 67 | |
| BX933650.2 | - | 6 - 7 | 513 | 67 | ||
| BX933652.1 | - | 6 - 7 | 513 | 67 | ||
| BX936227.2 | - | 6 - 7 | 513 | 67 | ||
| BX936267.2 | - | 6 - 7 | 513 | 67 | ||
| CR353143.1 | - | 6 - 7 | 501 | 67 | ||
More Information
Set Membership: |
|

PSMA4
Genomic Map