Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277828.1 | NP_001264757.1 | 8 - 9 | 648 | 62 | |
| XM_015276168.2 | XP_015131654.1 | 8 - 9 | 724 | 62 | ||
| XM_015276169.2 | XP_015131655.1 | 8 - 9 | 664 | 62 | ||
| XM_015276170.2 | XP_015131656.1 | 8 - 9 | 1090 | 62 | ||
| XM_015276171.2 | XP_015131657.1 | 8 - 9 | 2124 | 62 | ||
| XM_015276172.2 | XP_015131658.1 | 7 - 8 | 597 | 62 | ||
| GenBank mRNA | BX934030.1 | - | 8 - 9 | 645 | 62 | |
| CR406510.1 | - | 8 - 9 | 696 | 62 | ||
More Information
Set Membership: |
|

ZNF330
Genomic Map