Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642788.4 | XP_003642836.1 | 3 - 4 | 1637 | 68 | |
| GenBank mRNA | BX934585.1 | - | 3 - 4 | 818 | 68 | |
More Information
Set Membership: |
|

ZNF652
Genomic Map