Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418266.5 | XP_418266.2 | 3 - 4 | 294 | 64 | |
| GenBank mRNA | BX934951.1 | - | 3 - 4 | 294 | 64 | |
| CR353360.1 | - | 3 - 4 | 283 | 64 | ||
More Information
Set Membership: |
|

FAM32A
Genomic Map