Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940673.3 | XP_004940730.1 | 3 - 4 | 435 | 64 | |
| XM_015278476.2 | XP_015133962.1 | 3 - 4 | 479 | 64 | ||
| XM_420226.6 | XP_420226.2 | 3 - 4 | 431 | 64 | ||
| GenBank mRNA | BX935084.1 | - | 3 - 4 | 414 | 64 | |
| BX936027.2 | - | 3 - 4 | 409 | 64 | ||
More Information
Set Membership: |
|

MOSPD1
Genomic Map