Assay Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277811.2 | NP_001264740.1 | 5 - 6 | 1166 | 69 | |
| XM_015278141.2 | XP_015133627.1 | 6 - 7 | 1082 | 69 | ||
| XM_015278143.2 | XP_015133629.1 | 6 - 7 | 1215 | 69 | ||
| XM_015278147.2 | XP_015133633.1 | 5 - 6 | 1021 | 69 | ||
| GenBank mRNA | BX935095.1 | - | 5 - 6 | 1166 | 69 | |
| BX935779.2 | - | 5 - 6 | 886 | 69 | ||
More Information
Set Membership: |
|

PDGFD
Genomic Map