Assay Details
Gene Symbol: SGCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293900.2 | XP_015149386.1 | 4 - 5 | 663 | 68 | |
XM_015293901.2 | XP_015149387.1 | 4 - 5 | 542 | 68 | ||
XM_015293902.2 | XP_015149388.1 | 4 - 5 | 574 | 68 | ||
XM_015293903.2 | XP_015149389.1 | 4 - 5 | 559 | 68 | ||
XM_015293904.2 | XP_015149390.1 | 4 - 5 | 660 | 68 | ||
XM_015293905.2 | XP_015149391.1 | 3 - 4 | 439 | 68 | ||
XM_015293906.2 | XP_015149392.1 | 3 - 4 | 366 | 68 | ||
XM_015293908.2 | XP_015149394.1 | 4 - 5 | 666 | 68 | ||
XM_015293909.2 | XP_015149395.1 | 4 - 5 | 665 | 68 | ||
XM_015293910.2 | XP_015149396.1 | 4 - 5 | 663 | 68 | ||
XM_025154986.1 | XP_025010754.1 | 4 - 5 | 543 | 68 | ||
XM_025154987.1 | XP_025010755.1 | 4 - 5 | 666 | 68 | ||
XM_025154988.1 | XP_025010756.1 | 4 - 5 | 666 | 68 | ||
GenBank mRNA | BX950245.1 | - | 4 - 5 | 531 | 68 | |
CR405962.1 | - | 3 - 4 | 424 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
