Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277839.2 | NP_001264768.1 | 4 - 5 | 867 | 91 | |
| XM_015297923.2 | XP_015153409.1 | 4 - 5 | 872 | 91 | ||
| XM_015297924.2 | XP_015153410.1 | 4 - 5 | 1430 | 91 | ||
| XM_015297925.2 | XP_015153411.1 | 4 - 5 | 1391 | 91 | ||
| XM_015297926.2 | XP_015153412.1 | 4 - 5 | 1408 | 91 | ||
| XM_015297927.2 | XP_015153413.1 | 3 - 4 | 1276 | 91 | ||
| XM_015297928.2 | XP_015153414.1 | 3 - 4 | 1239 | 91 | ||
| GenBank mRNA | AF468195.1 | - | 1 - 2 | 151 | 91 | |
| AJ567375.1 | - | 4 - 5 | 867 | 91 | ||
| BX931547.2 | - | 5 - 6 | 768 | 91 | ||
More Information
Set Membership: |
|

B3GAT1
Genomic Map