Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419413.5 | XP_419413.1 | 1 - 2 | 94 | ||
| GenBank mRNA | BX932174.2 | - | 1 - 2 | 73 | 94 | |
| BX932335.2 | - | 1 - 2 | 76 | 94 | ||
| BX936075.1 | - | 1 - 2 | 76 | 94 | ||
More Information
Set Membership: |
|

RRP15
Genomic Map