Assay Details
| Gene Symbol: BATF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419428.6 | XP_419428.4 | 1 - 2 | 713 | 85 | |
| GenBank mRNA | BX932427.2 | - | 1 - 2 | 112 | 85 | |
| BX932508.1 | - | 1 - 2 | 110 | 85 | ||
More Information
Set Membership: |
|

BATF3
Genomic Map