Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277670.1 | NP_001264599.1 | 1 - 2 | 157 | 67 | |
| XM_025149572.1 | XP_025005340.1 | 4 - 5 | 1586 | 67 | ||
| GenBank mRNA | BX933547.2 | - | 1 - 2 | 117 | 67 | |
| CR354354.1 | - | 1 - 2 | 169 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277670.1, XM_025149572.1 transcripts.
Set Membership: |
|

ASB12
Genomic Map