Assay Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277580.1 | NP_001264509.1 | 2 - 3 | 188 | 68 | |
| XM_015291002.2 | XP_015146488.1 | 2 - 3 | 190 | 68 | ||
| GenBank mRNA | BX933138.2 | - | 2 - 3 | 174 | 68 | |
| BX933889.2 | - | 2 - 3 | 151 | 68 | ||
| BX934725.1 | - | 2 - 3 | 182 | 68 | ||
| CN233827.1 | - | 2 - 3 | 188 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015291002.2 transcripts.
Set Membership: |
|

TMEM53
Genomic Map