Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199519.1 | NP_001186448.1 | 5 - 6 | 766 | 66 | |
| GenBank mRNA | BX933611.1 | - | 5 - 6 | 766 | 66 | |
| BX935026.2 | - | 5 - 6 | 666 | 66 | ||
| BX935281.1 | - | 5 - 6 | 766 | 66 | ||
| BX935287.2 | - | 5 - 6 | 766 | 66 | ||
More Information
Set Membership: |
|

MAT1A
Genomic Map