Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198650.1 | NP_001185579.1 | 1 - 2 | 42 | 67 | |
| GenBank mRNA | BX934957.1 | - | 1 - 2 | 42 | 67 | |
| BX935785.2 | - | 1 - 2 | 30 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198650.1 transcripts.
Set Membership: |
|

MRPS33
Genomic Map