Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151304.1 | XP_025007072.1 | 2 - 3 | 243 | 61 | |
| GenBank mRNA | BX930048.1 | - | 1 - 2 | 46 | 61 | |
| BX932516.1 | - | 1 - 2 | 42 | 61 | ||
| BX936079.2 | - | 1 - 2 | 59 | 61 | ||
| CR385483.1 | - | 1 - 2 | 64 | 61 | ||
| CR407467.1 | - | 1 - 2 | 63 | 61 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map