Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198857.2 | NP_001185786.1 | 6 | 1484 | 127 | |
| GenBank mRNA | AJ727069.1 | - | 1 | 147 | 126 | |
| BU262072.1 | - | 1 | 438 | 125 | ||
| CR386344.1 | - | 1 | 956 | 126 | ||
More Information
Set Membership: |
|

CNEP1R1
Genomic Map