Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147420.1 | XP_025003188.1 | 7 - 8 | 6732 | 77 | |
| XM_025147421.1 | XP_025003189.1 | 7 - 8 | 6234 | 77 | ||
| XM_025147422.1 | XP_025003190.1 | 8 - 9 | 5512 | 77 | ||
| XM_025147423.1 | XP_025003191.1 | 8 - 9 | 5508 | 77 | ||
| XM_025147424.1 | XP_025003192.1 | 6 - 7 | 7920 | 77 | ||
| XM_418719.6 | XP_418719.1 | 6 - 7 | 7934 | 77 | ||
| GenBank mRNA | CR385674.1 | - | 1 - 2 | 68 | 77 | |
More Information
Set Membership: |
|

FAM221A
Genomic Map