Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX932823.1 | - | 7 - 8 | 760 | 76 | |
| CR385785.1 | - | 5 - 6 | 459 | 76 | ||
| CR386500.1 | - | 7 - 8 | 716 | 76 | ||
More Information
Set Membership: |
|

STOML2
Genomic Map