Assay Details
| Gene Symbol: TMEM174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282271.1 | NP_001269200.1 | 2 | 2816 | 75 | |
| XM_025144761.1 | XP_025000529.1 | 6 | 10172 | 75 | ||
| GenBank mRNA | CR388511.1 | - | 1 | 543 | 75 | |
More Information
Set Membership: |
|

TMEM174
Genomic Map