Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293095.1 | NP_001280024.1 | 8 - 9 | 1125 | 101 | |
| XM_004937021.3 | XP_004937078.3 | 9 - 10 | 1180 | 101 | ||
| XM_015276819.2 | XP_015132305.2 | 9 - 10 | 1180 | 101 | ||
| XM_025153314.1 | XP_025009082.1 | 9 - 10 | 1072 | 101 | ||
| GenBank mRNA | AF075706.1 | - | 2 - 3 | 150 | 101 | |
More Information
Set Membership: |
|

ETV5
Genomic Map