Assay Details
| Gene Symbol: FAM192A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277541.1 | NP_001264470.1 | 6 - 7 | 645 | 93 | |
| GenBank mRNA | BU120230.1 | - | 2 - 3 | 171 | 93 | |
| BX931924.2 | - | 2 - 3 | 668 | 93 | ||
| BX935399.1 | - | 6 - 7 | 630 | 93 | ||
| BX936029.2 | - | 6 - 7 | 622 | 93 | ||
More Information
Set Membership: |
|

FAM192A
Genomic Map