Assay Details
| Gene Symbol: CLDN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936975.3 | XP_004937032.3 | 4 - 5 | 628 | 67 | |
| XM_422589.4 | XP_422589.2 | 5 - 6 | 763 | 67 | ||
| GenBank mRNA | CR406782.1 | - | 5 - 6 | 687 | 67 | |
More Information
Set Membership: |
|

CLDN15
Genomic Map