Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282412.2 | XP_015137898.1 | 10 | 2663 | 110 | |
| XM_416030.6 | XP_416030.4 | 8 | 1931 | 110 | ||
| GenBank mRNA | CR407455.1 | - | 1 | 448 | 110 | |
More Information
Set Membership: |
|

PDZRN4
Genomic Map