Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279837.1 | XP_015135323.1 | 7 - 8 | 940 | 73 | |
| XM_015279838.2 | XP_015135324.1 | 7 - 8 | 940 | 73 | ||
| GenBank mRNA | DQ082894.1 | - | 5 - 6 | 666 | 73 | |
More Information
Set Membership: |
|

LHX6
Genomic Map