Assay Details
| Gene Symbol: ALPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205360.1 | NP_990691.1 | 6 - 7 | 839 | 55 | |
| GenBank mRNA | U19108.1 | - | 6 - 7 | 839 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALPL| Gene Symbol: ALPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205360.1 | NP_990691.1 | 6 - 7 | 839 | 55 | |
| GenBank mRNA | U19108.1 | - | 6 - 7 | 839 | 55 | |
Set Membership: |
|