Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293287.1 | NP_001280216.1 | 2 - 3 | 601 | 85 | |
| XM_004937989.3 | XP_004938046.1 | 2 - 3 | 761 | 85 | ||
| GenBank mRNA | AM068240.1 | - | 2 - 3 | 495 | 85 | |
| U92977.1 | - | 1 - 2 | 325 | 85 | ||
More Information
Set Membership: |
|

KCNJ8
Genomic Map