Assay Details
| Gene Symbol: MARCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286368.2 | NP_001273297.1 | 6 - 7 | 802 | 82 | |
| NM_001286369.2 | NP_001273298.1 | 6 - 7 | 1484 | 82 | ||
| XM_015288379.2 | XP_015143865.1 | 8 - 9 | 1827 | 82 | ||
| XM_015288380.2 | XP_015143866.1 | 8 - 9 | 1717 | 82 | ||
| XM_015288381.2 | XP_015143867.1 | 8 - 9 | 1768 | 82 | ||
| XM_015288382.2 | XP_015143868.1 | 8 - 9 | 1637 | 82 | ||
| XM_015288384.2 | XP_015143870.1 | 7 - 8 | 1693 | 82 | ||
| XM_025151577.1 | XP_025007345.1 | 9 - 10 | 2229 | 82 | ||
| XM_025151578.1 | XP_025007346.1 | 9 - 10 | 2174 | 82 | ||
| XM_025151579.1 | XP_025007347.1 | 8 - 9 | 1831 | 82 | ||
| XM_025151580.1 | XP_025007348.1 | 8 - 9 | 1667 | 82 | ||
| XM_025151581.1 | XP_025007349.1 | 7 - 8 | 1646 | 82 | ||
| XM_025151582.1 | XP_025007350.1 | 6 - 7 | 1643 | 82 | ||
| XM_025151583.1 | XP_025007351.1 | 6 - 7 | 817 | 82 | ||
| XM_025151584.1 | XP_025007352.1 | 6 - 7 | 745 | 82 | ||
| GenBank mRNA | BX931503.2 | - | 6 - 7 | 802 | 82 | |
| BX931843.1 | - | 5 - 6 | 654 | 82 | ||
More Information
Set Membership: |
|

MARCH8
Genomic Map