Assay Details
| Gene Symbol: LOC422609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276477.2 | XP_015131963.1 | 4 - 5 | 620 | 97 | |
| XM_015276478.2 | XP_015131964.1 | 3 - 4 | 5972 | 97 | ||
| XM_025150504.1 | XP_025006272.1 | 4 - 5 | 543 | 97 | ||
| XM_025150505.1 | XP_025006273.1 | 2 - 3 | 960 | 97 | ||
| XM_420568.5 | XP_420568.2 | 3 - 4 | 688 | 97 | ||
| GenBank mRNA | BX932483.1 | - | 1 - 2 | 200 | 97 | |
More Information
Set Membership: |
|

LOC422609
Genomic Map