Assay Details
| Gene Symbol: MCEE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293217.1 | NP_001280146.1 | 2 - 3 | 468 | 70 | |
| NM_001293218.1 | NP_001280147.1 | 2 - 3 | 429 | 70 | ||
| GenBank mRNA | BX266728.4 | - | 2 - 3 | 410 | 70 | |
| BX930043.2 | - | 2 - 3 | 429 | 70 | ||
| BX933690.1 | - | 2 - 3 | 374 | 70 | ||
| BX936254.1 | - | 2 - 3 | 374 | 70 | ||
More Information
Set Membership: |
|

MCEE
Genomic Map