Assay Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152394.1 | XP_025008162.1 | 2 - 3 | 345 | 58 | |
| GenBank mRNA | BX936098.2 | - | 2 - 3 | 263 | 58 | |
More Information
Set Membership: |
|

RAMP1
Genomic Map