Assay Details
Gene Symbol: BANP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173545.1 | NP_001167016.1 | 8 - 9 | 1209 | 62 | |
NM_001173546.1 | NP_001167017.1 | 8 - 9 | 1209 | 62 | ||
XM_015292561.2 | XP_015148047.1 | 9 - 10 | 1357 | 62 | ||
XM_015292562.2 | XP_015148048.1 | 10 - 11 | 1377 | 62 | ||
XM_015292563.2 | XP_015148049.1 | 9 - 10 | 1357 | 62 | ||
XM_015292564.2 | XP_015148050.1 | 9 - 10 | 1357 | 62 | ||
XM_015292565.2 | XP_015148051.1 | 9 - 10 | 1357 | 62 | ||
XM_015292566.2 | XP_015148052.1 | 8 - 9 | 1285 | 62 | ||
XM_015292567.2 | XP_015148053.1 | 9 - 10 | 1357 | 62 | ||
XM_025154185.1 | XP_025009953.1 | 9 - 10 | 1403 | 62 | ||
GenBank mRNA | BX950316.1 | - | 8 - 9 | 1209 | 62 | |
BX950441.2 | - | 3 - 4 | 493 | 62 | ||
CR406239.1 | - | 4 - 5 | 632 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
