Assay Details
| Gene Symbol: MANEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419831.6 | XP_419831.2 | 2 - 3 | 676 | 71 | |
| GenBank mRNA | CR354351.1 | - | 2 - 3 | 659 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MANEA| Gene Symbol: MANEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419831.6 | XP_419831.2 | 2 - 3 | 676 | 71 | |
| GenBank mRNA | CR354351.1 | - | 2 - 3 | 659 | 71 | |
Set Membership: |
|