Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277371.1 | NP_001264300.1 | 5 - 6 | 546 | 77 | |
| GenBank mRNA | CR387675.1 | - | 5 - 6 | 533 | 77 | |
| CR389655.1 | - | 1 - 2 | 68 | 77 | ||
More Information
Set Membership: |
|

RRP7A
Genomic Map