Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290547.1 | NP_001277476.1 | 2 - 3 | 501 | 132 | |
| XM_025153310.1 | XP_025009078.1 | 3 - 4 | 2508 | 132 | ||
| GenBank mRNA | AF217225.1 | - | 1 - 2 | 50 | 132 | |
| BX934658.1 | - | 1 - 2 | 100 | 132 | ||
| JN412068.1 | - | 2 - 3 | 501 | 132 | ||
More Information
Set Membership: |
|

HTR2B
Genomic Map