Assay Details
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004948137.3 | XP_004948194.1 | 5 - 6 | 804 | 66 | |
XM_015298297.2 | XP_015153783.1 | 5 - 6 | 744 | 66 | ||
XM_015298299.2 | XP_015153785.1 | 4 - 5 | 437 | 66 | ||
XM_417928.6 | XP_417928.2 | 5 - 6 | 389 | 66 | ||
GenBank mRNA | AF328139.1 | - | 1 - 2 | 47 | 66 | |
BX929645.1 | - | 4 - 5 | 580 | 66 | ||
BX931275.1 | - | 2 - 3 | 485 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
