Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031287.2 | NP_001026458.2 | 4 - 5 | 1646 | 62 | |
| XM_025152880.1 | XP_025008648.1 | 4 - 5 | 1765 | 62 | ||
| GenBank mRNA | AJ720201.1 | - | 4 - 5 | 1646 | 62 | |
More Information
Set Membership: |
|

CPT2
Genomic Map